missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Hydroxyhexanoate 98.0+%, TCI America™

Catalog No. E078825G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E078825G 25 g
E0788100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E078825G Supplier TCI America Supplier No. E078825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2305-25-1
Molecular Formula C8H16O3
Molecular Weight (g/mol) 160.213
MDL Number MFCD00036604
InChI Key LYRIITRHDCNUHV-UHFFFAOYSA-N
Synonym hexanoic acid, 3-hydroxy-, ethyl ester, ethyl beta-hydroxycaproate, ethyl 3-hydroxy-hexanoate, ethyl hydroxy-3-hexanoate, 3-hydroxyhexanoic acid ethyl ester, fema no. 3545, acmc-20lakf, ethyl 3-hydroxycaproate, ethyl-3-hydrixyhexanoate, ethyl 3-hydroxy hexanoate
PubChem CID 61293
ChEBI CHEBI:23997
IUPAC Name ethyl 3-hydroxyhexanoate
SMILES CCCC(CC(=O)OCC)O

Specifications

Color Colorless
Boiling Point 92°C
Quantity 25 g
Formula Weight 160.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Hydroxyhexanoate
EINECSNumber (2)-1430
TSCA Yes