missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Furancarboxylate 98.0+%, TCI America™

Catalog No. F04635G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04635G 5 g
F046325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. F04635G missing translation for 'mfr' TCI America missing translation for 'supplierNo' F04635G
Only null left

Chemical Identifiers

CAS 614-98-2
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00005348
InChI Key LOFDXZJSDVCYAS-UHFFFAOYSA-N
PubChem CID 69201
IUPAC Name ethyl furan-3-carboxylate
SMILES CCOC(=O)C1=COC=C1

Specifications

Color Yellow
Boiling Point 95°C
Quantity 5 g
UN Number 3272
Synonym ethyl 3-furoate, ethyl 3-furancarboxylate, unii-i8q0yn919k, 3-furancarboxylic acid, ethyl ester, 3-furancarboxylic acid ethyl ester, ethyl 3furoate, ethyl3-furancarboxylate, ethyl3-furoate, pubchem19999, acmc-1bczk
Formula Weight 140.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Furancarboxylate
TSCA No
Recommended Storage Refrigerator