missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Ethoxyacrylate (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. E00415ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E00415ML 5 mL
E004125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E00415ML Supplier TCI America Supplier No. E00415ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1001-26-9
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00009863
InChI Key ITQFPVUDTFABDH-AATRIKPKSA-N
PubChem CID 5366269
IUPAC Name ethyl (E)-3-ethoxyprop-2-enoate
SMILES CCOC=CC(=O)OCC

Specifications

Color Yellow
Quantity 5 mL
Synonym ethyl 3-ethoxyacrylate, e-ethyl 3-ethoxyacrylate, ethyl 3-ethoxy-2-propenoate, ethyl trans-3-ethoxyacrylate, ethyl .beta.-ethoxyacrylate, 3-ethoxyacrylic acid ethyl ester, ethyl 2e-3-ethoxyprop-2-enoate, e-3-ethoxy-acrylic acid ethyl ester, ethyl3-ethoxyacrylate, ethyl 3-ethoxypropenoate
Formula Weight 144.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Ethoxyacrylate (cis- and trans- mixture)