missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 3-Chlorobenzoate 94.0+%, TCI America™
Supplier: TCI America C173725G
Specifications
| Ethyl 3-Chlorobenzoate | |
| Yellow | |
| MFCD00013634 | |
| benzoic acid, 3-chloro-, ethyl ester, 3-chlorobenzoic acid ethyl ester, ethyl m-chlorobenzoate, benzoic acid, m-chloro-, ethyl ester, 3-chloro-benzoic acid ethyl ester, pubchem3704, acmc-1bt4u, ethyl3-chlorobenzoate, ksc495m6f, rarechem al bi 0056 | |
| CCOC(=O)C1=CC(=CC=C1)Cl | |
| 184.619 | |
| 184.62 | |
| Liquid |
| 1128-76-3 | |
| C9H9ClO2 | |
| 25 g | |
| LVFRSNCBCHABAM-UHFFFAOYSA-N | |
| ethyl 3-chlorobenzoate | |
| 70785 | |
| ≥94.0% (GC) |
Chemical Identifiers
| 1128-76-3 | |
| 184.619 | |
| LVFRSNCBCHABAM-UHFFFAOYSA-N | |
| 70785 | |
| CCOC(=O)C1=CC(=CC=C1)Cl |
| C9H9ClO2 | |
| MFCD00013634 | |
| benzoic acid, 3-chloro-, ethyl ester, 3-chlorobenzoic acid ethyl ester, ethyl m-chlorobenzoate, benzoic acid, m-chloro-, ethyl ester, 3-chloro-benzoic acid ethyl ester, pubchem3704, acmc-1bt4u, ethyl3-chlorobenzoate, ksc495m6f, rarechem al bi 0056 | |
| ethyl 3-chlorobenzoate |
Safety and Handling
TSCA : No