missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Chlorobenzoate 94.0+%, TCI America™

Catalog No. p-7125145
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
C17375G 5 g
C173725G 25 g
2 options

Catalog No. C17375G

Supplier: TCI America C17375G

Only null left

Chemical Identifiers

CAS 1128-76-3
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00013634
InChI Key LVFRSNCBCHABAM-UHFFFAOYSA-N
Synonym benzoic acid, 3-chloro-, ethyl ester, 3-chlorobenzoic acid ethyl ester, ethyl m-chlorobenzoate, benzoic acid, m-chloro-, ethyl ester, 3-chloro-benzoic acid ethyl ester, pubchem3704, acmc-1bt4u, ethyl3-chlorobenzoate, ksc495m6f, rarechem al bi 0056
PubChem CID 70785
IUPAC Name ethyl 3-chlorobenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)Cl

Specifications

Color Yellow
Quantity 5 g
Formula Weight 184.62
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Chlorobenzoate