accessibility menu, dialog, popup

UserName

CAS RN 1128-76-3

IUPAC Name: ethyl 3-chlorobenzoate
Synonyms: ethyl 3-chlorobenzoate
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: LVFRSNCBCHABAM-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC(Cl)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)