missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-bromobenzoate, 98%, Thermo Scientific™

Numéro de catalogue. AC308540250
Modifier l'affichage
Click to view available options
Quantité:
5g
25g
Emballage:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC308540250 25g Glass bottle
AC308540050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC308540250 Fournisseur Thermo Scientific Chemicals Code fournisseur 308540250

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 24398-88-7
Formule moléculaire C9H9BrO2
Poids moléculaire (g/mol) 229.08
Numéro MDL MFCD00013529
Clé InChI QAUASTLEZAPQTB-UHFFFAOYSA-N
Synonyme ethyl3-bromobenzoate, 3-bromobenzoic acid ethyl ester, benzoic acid, 3-bromo-, ethyl ester, ethyl-3-bromobenzoate, benzoic acid, m-bromo-, ethyl ester, m-bromo benzoic acid ethyl ester, m-bromobenzoic acid, ethyl ester, pubchem3941, acmc-209gbv, ethyl 3-bromobenzoate
PubChem CID 90488
Nom de l’IUPAC ethyl 3-bromobenzoate
SOURIRES CCOC(=O)C1=CC(=CC=C1)Br

Spécifications

Nom ou substance chimique Ethyl 3-bromobenzoate
Densité 1.4300g/mL
Point d’ébullition 130°C to 131°C (12.0mmHg)
Point d’éclair >110°C
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 97.5% min. (GC)
Emballage Glass bottle
Formule linéaire BrC6H4CO2C2H5
Indice de réfraction 1.5405 to 1.5425
Quantité 25g
Gravité spécifique 1.43
Poids de formule 229.08
Pourcentage de pureté 98%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 246-223-9

RUO – Research Use Only