Learn More
Ethyl 3-aminobenzoate, 98%
CAS: 582-33-2 | C9H11NO2 | 165.192 g/mol
$70.62 - $1223.36
Chemical Identifiers
| CAS | 582-33-2 |
|---|---|
| Molecular Formula | C9H11NO2 |
| Molecular Weight (g/mol) | 165.192 |
| MDL Number | MFCD00007794 |
| InChI Key | ZMCBYSBVJIMENC-UHFFFAOYSA-N |
| Synonym | 3-aminobenzoic acid ethyl ester, m-ethoxycarbonylaniline, 3-ethoxycarbonyl aniline, ethyl m-aminobenzoate, benzoic acid, 3-amino-, ethyl ester, m-aminobenzoic acid, ethyl ester, m-aminobenzoate, metacaine, 3-amino-benzoic acid ethyl ester, benzoic acid, m-amino-, ethyl ester |
| PubChem CID | 11400 |
| ChEBI | CHEBI:81494 |
| IUPAC Name | ethyl 3-aminobenzoate |
| SMILES | CCOC(=O)C1=CC(=CC=C1)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1444809
|
Thermo Scientific Chemicals
A1444809 |
10 g |
Each for $70.62
|
|
|||||
|
AAA1444818
|
Thermo Scientific Chemicals
A1444818 |
50 g |
Each for $278.92
|
|
|||||
|
AAA1444822
|
Thermo Scientific Chemicals
A1444822 |
100 g |
Each for $510.18
|
|
|||||
|
AAA1444830
|
Thermo Scientific Chemicals
A1444830 |
250 g |
Each for $1,223.36
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 582-33-2 | |
| 165.192 | |
| ZMCBYSBVJIMENC-UHFFFAOYSA-N | |
| 11400 | |
| ethyl 3-aminobenzoate |
| C9H11NO2 | |
| MFCD00007794 | |
| 3-aminobenzoic acid ethyl ester, m-ethoxycarbonylaniline, 3-ethoxycarbonyl aniline, ethyl m-aminobenzoate, benzoic acid, 3-amino-, ethyl ester, m-aminobenzoic acid, ethyl ester, m-aminobenzoate, metacaine, 3-amino-benzoic acid ethyl ester, benzoic acid, m-amino-, ethyl ester | |
| CHEBI:81494 | |
| CCOC(=O)C1=CC(=CC=C1)N |
Specifications
| 582-33-2 | |
| 1.106 | |
| >110°C (230°F) | |
| 1.561 | |
| 10 g | |
| 3-aminobenzoic acid ethyl ester, m-ethoxycarbonylaniline, 3-ethoxycarbonyl aniline, ethyl m-aminobenzoate, benzoic acid, 3-amino-, ethyl ester, m-aminobenzoic acid, ethyl ester, m-aminobenzoate, metacaine, 3-amino-benzoic acid ethyl ester, benzoic acid, m-amino-, ethyl ester | |
| CCOC(=O)C1=CC(=CC=C1)N | |
| 165.192 | |
| CHEBI:81494 | |
| 98% |
| 27°C to 30°C | |
| 294°C | |
| C9H11NO2 | |
| MFCD00007794 | |
| 1101967 | |
| ZMCBYSBVJIMENC-UHFFFAOYSA-N | |
| ethyl 3-aminobenzoate | |
| 11400 | |
| 165.19 | |
| Ethyl 3-aminobenzoate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 209-482-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only