accessibility menu, dialog, popup

UserName

H3COOH2N

CAS RN 582-33-2

IUPAC Name: ethyl 3-aminobenzoate
Synonyms: tricaine
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: ZMCBYSBVJIMENC-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC(N)=C1
Boiling Point 
  • (2)
  • (5)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (1)
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)