missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-aminobenzoate, 98%

CAS: 582-33-2 | C9H11NO2 | 165.192 g/mol

Supplier:  Thermo Scientific Chemicals A1444830

 View more versions of this product

Catalog No. AAA1444830


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl 3-aminobenzoate
27°C to 30°C
294°C
C9H11NO2
MFCD00007794
1101967
ZMCBYSBVJIMENC-UHFFFAOYSA-N
ethyl 3-aminobenzoate
11400
165.19
582-33-2
1.106
>110°C (230°F)
1.561
250 g
3-aminobenzoic acid ethyl ester, m-ethoxycarbonylaniline, 3-ethoxycarbonyl aniline, ethyl m-aminobenzoate, benzoic acid, 3-amino-, ethyl ester, m-aminobenzoic acid, ethyl ester, m-aminobenzoate, metacaine, 3-amino-benzoic acid ethyl ester, benzoic acid, m-amino-, ethyl ester
CCOC(=O)C1=CC(=CC=C1)N
165.192
CHEBI:81494
98%

Chemical Identifiers

582-33-2
165.192
ZMCBYSBVJIMENC-UHFFFAOYSA-N
11400
ethyl 3-aminobenzoate
C9H11NO2
MFCD00007794
3-aminobenzoic acid ethyl ester, m-ethoxycarbonylaniline, 3-ethoxycarbonyl aniline, ethyl m-aminobenzoate, benzoic acid, 3-amino-, ethyl ester, m-aminobenzoic acid, ethyl ester, m-aminobenzoate, metacaine, 3-amino-benzoic acid ethyl ester, benzoic acid, m-amino-, ethyl ester
CHEBI:81494
CCOC(=O)C1=CC(=CC=C1)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 209-482-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only