missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Amino-4,4,4-trifluorocrotonate 97.0+%, TCI America™

Catalog No. A14755G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14755G 5 g
A147525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14755G Supplier TCI America Supplier No. A14755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 372-29-2
Molecular Formula C6H8F3NO2
Molecular Weight (g/mol) 183.13
MDL Number MFCD00068195
InChI Key NXVKRKUGIINGHD-ONEGZZNKSA-N
PubChem CID 10899311
IUPAC Name ethyl (E)-3-amino-4,4,4-trifluorobut-2-enoate
SMILES CCOC(=O)C=C(C(F)(F)F)N

Specifications

Color White-Yellow
Boiling Point 83°C
Quantity 5 g
Synonym ethyl 3-amino-4,4,4-trifluorocrotonate, ethyl 3-amino-4,4,4-trifluorobut-2-enoate, ethyl 2z-3-amino-4,4,4-trifluorobut-2-enoate, 3-amino-4,4,4-trifluorocrotonic acid ethyl ester, ethyl-3-amino-4,4,4-trifluorocrotonate, ethyl 3-amino-4,4,4-trifluoro-2-butenoate, ethyl z-3-amino-4,4,4-trifluorobut-2-enoate, ethyl z-3-amino-4,4,4-trifluoro-but-2-enoate, hcvdpjfpdddldlbdlrfuulu@bh, ethyl3-amino-4,4,4-trifluorocrotonate
Formula Weight 183.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Ethyl 3-Amino-4,4,4-trifluorocrotonate