missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Amino-2-butenoate 98.0+%, TCI America™

Catalog No. E070925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E070925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E070925G Supplier TCI America Supplier No. E070925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7318-00-5
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD00008073
InChI Key YPMPTULBFPFSEQ-PLNGDYQASA-N
Synonym z-ethyl 3-aminobut-2-enoate, ethyl 3-aminocrotonate, ethyl 3-aminobut-2-enoate, ethyl 2z-3-aminobut-2-enoate, 2-butenoic acid, 3-amino-, ethyl ester, 2z, ethyl z-3-aminobut-2-enoate, ethyl 3-amino-2-butenoate, .beta.-aminocrotonic acid ethyl ester, 2-butenoic acid, 3-amino-, ethyl ester, ethyl .beta.-aminocrotonate
PubChem CID 643756
IUPAC Name ethyl (Z)-3-aminobut-2-enoate
SMILES CCOC(=O)C=C(C)N

Specifications

Color Yellow
Boiling Point 215°C
Quantity 25 g
Formula Weight 129.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Amino-2-butenoate