missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,5-Dichloro-4-hydroxybenzoate 98.0+%, TCI America™

Catalog No. D03745G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D03745G 5 g
D037425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D03745G Supplier TCI America Supplier No. D03745G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17302-82-8
Molecular Formula C9H8Cl2O3
Molecular Weight (g/mol) 235.06
MDL Number MFCD00016420
InChI Key WMKNGSJJEMFQOT-UHFFFAOYSA-N
Synonym 3,5-dichloro-4-hydroxybenzoic acid ethyl ester, benzoic acid, 3,5-dichloro-4-hydroxy-, ethyl ester, 3,5-dichloro-4-hydroxy-benzoic acid ethyl ester, acmc-209e5q, 3-10-00-00363 beilstein handbook reference, wln: qr bg fg dvo2, ethyl 3,5-dichloro-4-hydroxy-benzoate, ethyl 3,5-bis chloranyl-4-oxidanyl-benzoate, benzoic acid,5-dichloro-4-hydroxy-, ethyl ester
PubChem CID 28460
IUPAC Name ethyl 3,5-dichloro-4-hydroxybenzoate
SMILES CCOC(=O)C1=CC(=C(C(=C1)Cl)O)Cl

Specifications

Melting Point 112°C
Color White
Quantity 5 g
Formula Weight 235.06
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3,5-Dichloro-4-hydroxybenzoate
EINECSNumber (3)-4072
RTECSNumber DG7503500
TSCA No