missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(2-furyl)-1-methyl-1H-pyrazole-5-carboxylate, 97%, Thermo Scientific™

Numéro de catalogue. CC47023DA
Modifier l'affichage
Click to view available options
Quantité:
1g
Emballage:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC47023DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC47023DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC47023DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 104295-62-7
Formule moléculaire C11H12N2O3
Poids moléculaire (g/mol) 220.228
Numéro MDL MFCD08271951
Clé InChI RMIOHJSPVQALON-UHFFFAOYSA-N
Synonyme ethyl 3-2-furyl-1-methyl-1h-pyrazole-5-carboxylate, ethyl 5-furan-2-yl-2-methylpyrazole-3-carboxylate, 5-ethoxycarbonyl-3-2-furyl-1-methylpyrazole, 1h-pyrazole-5-carboxylicacid, 3-2-furanyl-1-methyl-, ethyl ester, acmc-1bsv1, ethyl 3-2-furyl-1-methylpyrazole-5-carboxylate, ethyl 3-furan-2-yl-1-methyl-1h-pyrazole-5-carboxylate, 1-methyl-3-2-furyl-1h-pyrazole-5-carboxylic acid ethyl ester, 1h-pyrazole-5-carboxylicacid,3-2-furanyl-1-methyl-,ethyl ester
PubChem CID 7537645
Nom de l’IUPAC ethyl 5-(furan-2-yl)-2-methylpyrazole-3-carboxylate
SOURIRES CCOC(=O)C1=CC(=NN1C)C2=CC=CO2

Spécifications

Nom ou substance chimique Ethyl 3-(2-furyl)-1-methyl-1H-pyrazole-5-carboxylate
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 1g
Poids de formule 220.23
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.