missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(2,6-Dichloro-5-fluoro-3-pyridyl)-3-oxopropionate 98.0+%, TCI America™

Catalog No. E08115G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08115G 5 g
E081125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E08115G Supplier TCI America Supplier No. E08115G

Chemical Identifiers

CAS 96568-04-6
Molecular Formula C10H8Cl2FNO3
Molecular Weight (g/mol) 280.08
MDL Number MFCD00799535
InChI Key IEUHWNLWVMLHHC-UHFFFAOYSA-N
Synonym ethyl 3-2,6-dichloro-5-fluoropyridin-3-yl-3-oxopropanoate, ethyl 3-2,6-dichloro-5-fluoro-3-pyridyl-3-oxopropionate, ethyl 2,6-dichloro-5-fluoronicotinoylacetate, ethyl 2,6-dichloro-5-fluoro pyridine-3-acetoacetate, ethyl 2,6-dichloro-5-fluoronicotinoyl acetate, ethyl 2,6-dichloro-5-fluornicotinoylacetate, ethyl 3-2,6-dichloro-5-fluoro-3-pyridiyl-3-oxopropanoate, 2,6-dichloro-5-fluoronicotinoyl acetic acid ethyl ester, ethyl 2,6-dichloro-5-fluoro nicotinylacetate, ethyl 3-2,6-dichloro-5-fluoro 3-pyridyl-3-oxopropanoate
PubChem CID 2733708
IUPAC Name ethyl 3-(2,6-dichloro-5-fluoropyridin-3-yl)-3-oxopropanoate
SMILES CCOC(=O)CC(=O)C1=CC(F)=C(Cl)N=C1Cl

Specifications

Melting Point 69°C
Quantity 5 g
Formula Weight 280.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3-(2,6-Dichloro-5-fluoro-3-pyridyl)-3-oxopropionate