missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(Triphenylphosphoranylidene)propionate 97.0+%, TCI America™

Catalog No. C16415G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C16415G Supplier TCI America Supplier No. C16415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5717-37-3
Molecular Formula C23H23O2P
Molecular Weight (g/mol) 362.41
MDL Number MFCD00009160
InChI Key KZENFXVDPUMQOE-UHFFFAOYSA-N
Synonym ethyl 2-triphenylphosphoranylidene propanoate, carbethoxyethylidene triphenylphosphorane, ethyl 2-triphenylphosphoranylidene propionate, 1-ethoxycarbonylethylidene triphenylphosphorane, propanoic acid, 2-triphenylphosphoranylidene-, ethyl ester, 1-ethoxycarbonyl ethylidenetriphenylphosphorane, ethyl 2-triphenyl-lambda5-phosphanylidene propanoate, ethyl 2-triphenyl-$l^ 5-phosphanylidene propanoate, 1-ethoxycarbonylethylethylidene triphenylphosporane, carbethoxyethylidine triphenylphosphorane
PubChem CID 79792
IUPAC Name ethyl 2-(triphenyl-λ⁵-phosphanylidene)propanoate
SMILES CCOC(=O)C(C)=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 161°C
Color Yellow
Quantity 5 g
Formula Weight 362.41
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-(Triphenylphosphoranylidene)propionate
TSCA No
Recommended Storage Freezer