missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-morpholinobenzoate, 97%, Thermo Scientific™

Numéro de catalogue. p-3175165
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC24323CB 250mg
CC24323DA 1g
CC24323EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. CC24323CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC24323CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 192817-79-1
Formule moléculaire C13H17NO3
Poids moléculaire (g/mol) 235.283
Numéro MDL MFCD06204497
Clé InChI UIVDSGDHUXOYDW-UHFFFAOYSA-N
Synonyme ethyl 2-morpholinobenzoate, ethyl 2-morpholin-4-yl benzoate, 2-morpholin-4-yl-benzoic acid ethyl ester, benzoic acid,2-4-morpholinyl-,ethyl ester, 2-morpholin-4-ylbenzoic acid ethyl ester, benzoic acid,2-4-morpholinyl-, ethyl ester
PubChem CID 7148401
Nom de l’IUPAC ethyl 2-morpholin-4-ylbenzoate
SOURIRES CCOC(=O)C1=CC=CC=C1N2CCOCC2

Spécifications

Nom ou substance chimique Ethyl 2-morpholinobenzoate
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber glass bottle
Quantité 250mg
Poids de formule 235.28
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.