missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-methylnicotinate, 97%

Catalog No. AAL0325614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0325614 25 g
AAL0325606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0325614 Supplier Thermo Scientific Chemicals Supplier No. L0325614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1721-26-2 | C9H11NO2 | 165.19 g/mol

Ethyl 2-methylpyridine-3-carboxylate was used in the preparation of amino esters such as ethyl 2(piperidinylaminomethyl)benzoate, ethyl 2-(N-benzyl-N-methylaminomethyl)benzoate, ethyl 2-(diethylaminomethyl)benzoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-methylpyridine-3-carboxylate was used in the preparation of amino esters such as ethyl 2(piperidinylaminomethyl)benzoate, ethyl 2-(N-benzyl-N-methylaminomethyl)benzoate, ethyl 2-(diethylaminomethyl)benzoate.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 1721-26-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00006336
InChI Key TZORNSWZUZDUCU-UHFFFAOYSA-N
Synonym ethyl 2-methylnicotinate, 2-methylnicotinic acid ethyl ester, 2-methyl-nicotinic acid ethyl ester, 3-pyridinecarboxylic acid, 2-methyl-, ethyl ester, ethyl 2-methyl nicotinate, ethyl 2-methyl-3-pyridinecarboxylate, 2-methyl-3-pyridinecarboxylic acid ethyl ester, 2-methylpyridine-3-carboxylic acid ethyl ester, ethyl 2-methylpyridine-3-carboxylate∼2-methylnicotinic acid ethyl ester, pubchem12727
PubChem CID 74401
IUPAC Name ethyl 2-methylpyridine-3-carboxylate
SMILES CCOC(=O)C1=CC=CN=C1C

Specifications

Density 1.07
Boiling Point 107°C to 108°C (13 mmHg)
Flash Point 102°C (215°F)
Odor Recognizable
Refractive Index 1.505
Quantity 25 g
Beilstein 118200
Solubility Information Slightly soluble in water.
Formula Weight 165.19
Percent Purity 97%
Chemical Name or Material Ethyl 2-methylnicotinate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-013-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only