missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-methylbutyrate, 98%

Catalog No. AAL06743AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL06743AP 500 mL
AAL06743AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL06743AP Supplier Thermo Scientific Chemicals Supplier No. L06743AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7452-79-1 | C7H14O2 | 130.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7452-79-1
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00012217
InChI Key HCRBXQFHJMCTLF-UHFFFAOYSA-N
Synonym ethyl 2-methylbutyrate, butanoic acid, 2-methyl-, ethyl ester, butyric acid, 2-methyl-, ethyl ester, ethyl 2-methyl butyrate, ethyl alpha-methylbutyrate, dl-2-methylbutyric acid ethyl ester, ethyl 2-methyl-butanoate, ethyl .alpha.-methylbutyrate, 2-methylbutanoic acid ethyl ester, butyric acid, 2-methyl-, ethyl ester 8ci
PubChem CID 24020
IUPAC Name ethyl 2-methylbutanoate
SMILES CCC(C)C(=O)OCC

Specifications

Density 0.869
Boiling Point 132°C to 133°C
Flash Point 26°C (78°F)
Refractive Index 1.397
Quantity 500 mL
UN Number UN3272
Beilstein 1720887
Formula Weight 130.19
Percent Purity 98%
Chemical Name or Material Ethyl 2-methylbutyrate
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 231-225-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only