missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-methylacetoacetate, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 326111000

Quantity 100g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC326111000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

609-14-3
144.17
FNENWZWNOPCZGK-UHFFFAOYSA-N
701
CCOC(=O)C(C)C(=O)C
C7H12O3
MFCD00009164
ethyl 2-methylacetoacetate, ethyl 2-acetylpropionate, ethyl-2-methyl acetoacetate, ethyl methylacetoacetate, ethyl 2-methyl-3-oxobutyrate, ethyl2-methylacetoacetate, butanoic acid, 2-methyl-3-oxo-, ethyl ester, 2-methylacetoacetic acid ethyl ester, ethyl alpha-methylacetylacetate, 2-methyl-3-oxobutanoic acid ethyl ester
ethyl 2-methyl-3-oxobutanoate

Specifications

Ethyl 2-methylacetoacetate
609-14-3
187°C
Authentic
Glass bottle
CH3COCH(CH3)CO2C2H5
100g
03, 679
ethyl 2-methylacetoacetate, ethyl 2-acetylpropionate, ethyl-2-methyl acetoacetate, ethyl methylacetoacetate, ethyl 2-methyl-3-oxobutyrate, ethyl2-methylacetoacetate, butanoic acid, 2-methyl-3-oxo-, ethyl ester, 2-methylacetoacetic acid ethyl ester, ethyl alpha-methylacetylacetate, 2-methyl-3-oxobutanoic acid ethyl ester
FNENWZWNOPCZGK-UHFFFAOYSA-N
ethyl 2-methyl-3-oxobutanoate
701
95%
95%
1.0100g/mL
62°C
94% min. (GC)
C7H12O3
1.4170 to 1.419
MFCD00009164
1.01
Solubility in water: insoluble. Other solubilities: miscible with most organic solvents
CCOC(=O)C(C)C(=O)C
144.17
144.17
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 210-179-9

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...