missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (2-Methoxyphenoxy)acetate 98.0+%, TCI America™

Catalog No. E12415G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12415G 5 g
E124125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12415G Supplier TCI America Supplier No. E12415G
Only null left

Chemical Identifiers

CAS 13078-21-2
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.229
MDL Number MFCD00082657
InChI Key BZCGGCRVJFWCIW-UHFFFAOYSA-N
Synonym (2-Methoxyphenoxy)acetic Acid Ethyl Ester
PubChem CID 83114
IUPAC Name ethyl 2-(2-methoxyphenoxy)acetate
SMILES CCOC(=O)COC1=CC=CC=C1OC

Specifications

Color Yellow
Boiling Point 116°C
Quantity 5 g
Formula Weight 210.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl (2-Methoxyphenoxy)acetate