missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Cyano-2-methylpropionate 98.0+%, TCI America™

Catalog No. C15465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15465G 5 g
C154625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15465G Supplier TCI America Supplier No. C15465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1572-98-1
Molecular Formula C7H11NO2
Molecular Weight (g/mol) 141.17
MDL Number MFCD00034667
InChI Key FYGRPGOHQCPZCV-UHFFFAOYSA-N
PubChem CID 344887
IUPAC Name ethyl 2-cyano-2-methylpropanoate
SMILES CCOC(=O)C(C)(C)C#N

Specifications

Color Colorless
Boiling Point 185°C
Quantity 5 g
Synonym 2-cyano-2-methylpropionic acid ethyl ester, ethyl 2-cyano-2-methylpropionate, ethyl 2-cyano-2,2-dimethylacetate, 2-cyano-2-methylpropanoic acid ethyl ester, ethyl 2-cyano-2-methyl-propanoate, propanoic acid, 2-cyano-2-methyl-, ethyl ester, pubchem23352, acmc-209dfv, ksc529g3h, ethyl2-cyano-2-methylpropionate
Formula Weight 141.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2-Cyano-2-methylpropionate