missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-Chloro-2-(hydroxyimino)acetate 99.0+%, TCI America™
Supplier: TCI America E037125G
Specifications
| Ethyl 2-Chloro-2-(hydroxyimino)acetate | |
| 81°C | |
| C4H6ClNO3 | |
| 25 g | |
| UXOLDCOJRAMLTQ-UTCJRWHESA-N | |
| ethyl (2Z)-2-chloro-2-(hydroxyimino)acetate | |
| 9576005 | |
| ≥99.0% (T) |
| 14337-43-0 | |
| White-Yellow | |
| MFCD00010209 | |
| ethyl 2-chloro-2-hydroxyimino acetate, 2-chloro-2-hydroxyiminoacetic acid ethyl ester, ethyl chlorooximinoacetate, ethyl chloro oximido acetate, ethyl chlorooximidoacetate, z-ethyl 2-chloro-2-hydroxyimino acetate, chloro oxime, ethyl 2z-2-chloro-2-hydroxyimino acetate, e-ethyl 2-chloro-2-hydroxyimino acetate, ethyl 2z-chloro hydroxyimino ethanoate | |
| CCOC(=O)C(\Cl)=N\O | |
| 151.55 | |
| 151.55 | |
| Crystalline Powder |
Chemical Identifiers
| 14337-43-0 | |
| 151.55 | |
| UXOLDCOJRAMLTQ-UTCJRWHESA-N | |
| 9576005 | |
| CCOC(=O)C(\Cl)=N\O |
| C4H6ClNO3 | |
| MFCD00010209 | |
| ethyl 2-chloro-2-hydroxyimino acetate, 2-chloro-2-hydroxyiminoacetic acid ethyl ester, ethyl chlorooximinoacetate, ethyl chloro oximido acetate, ethyl chlorooximidoacetate, z-ethyl 2-chloro-2-hydroxyimino acetate, chloro oxime, ethyl 2z-2-chloro-2-hydroxyimino acetate, e-ethyl 2-chloro-2-hydroxyimino acetate, ethyl 2z-chloro hydroxyimino ethanoate | |
| ethyl (2Z)-2-chloro-2-(hydroxyimino)acetate |
Safety and Handling
TSCA : Yes