missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Chloro-2-(hydroxyimino)acetate 99.0+%, TCI America™

Catalog No. p-7128966
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E03715G 5 g
E037125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E03715G Supplier TCI America Supplier No. E03715G

Chemical Identifiers

CAS 14337-43-0
Molecular Formula C4H6ClNO3
Molecular Weight (g/mol) 151.55
MDL Number MFCD00010209
InChI Key UXOLDCOJRAMLTQ-UTCJRWHESA-N
Synonym ethyl 2-chloro-2-hydroxyimino acetate, 2-chloro-2-hydroxyiminoacetic acid ethyl ester, ethyl chlorooximinoacetate, ethyl chloro oximido acetate, ethyl chlorooximidoacetate, z-ethyl 2-chloro-2-hydroxyimino acetate, chloro oxime, ethyl 2z-2-chloro-2-hydroxyimino acetate, e-ethyl 2-chloro-2-hydroxyimino acetate, ethyl 2z-chloro hydroxyimino ethanoate
PubChem CID 9576005
IUPAC Name ethyl (2Z)-2-chloro-2-(hydroxyimino)acetate
SMILES CCOC(=O)C(\Cl)=N\O

Specifications

Melting Point 81°C
Color White-Yellow
Quantity 5 g
Formula Weight 151.55
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-Chloro-2-(hydroxyimino)acetate