accessibility menu, dialog, popup

UserName

CH3OOClNHO(Z)

CAS RN 14337-43-0

IUPAC Name: ethyl (2Z)-2-chloro-2-(hydroxyimino)acetate
Synonyms: ethyl (2Z)-2-chloro-2-(hydroxyimino)acetate
Molecular Weight (g/mol): 151.55
Molecular Formula: C4H6ClNO3
InChi Key: UXOLDCOJRAMLTQ-UTCJRWHESA-N
SMILES: CCOC(=O)C(\Cl)=N\O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results