missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromoisobutyrate, 98+%

Catalog No. AAB2278518
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2278518 50 g
AAB2278530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2278518 Supplier Thermo Scientific Chemicals Supplier No. B2278518

CAS: 600-00-0 | C6H11BrO2 | 195.06 g/mol

Ethyl 2-bromoisobutyrate is used as an Atom Transfer Radical Polymerization (ATRP) initiator that will generate a polymer with an ethyl end group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-bromoisobutyrate is used as an Atom Transfer Radical Polymerization (ATRP) initiator that will generate a polymer with an ethyl end group.

Solubility
Soluble in alcohol, ether. Insoluble in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents, acids, bases, reducing agents.

Chemical Identifiers

CAS 600-00-0
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.06
MDL Number MFCD00000123
InChI Key IOLQWGVDEFWYNP-UHFFFAOYSA-N
Synonym ethyl 2-bromoisobutyrate, ethyl 2-bromo-2-methylpropionate, ethyl alpha-bromoisobutyrate, ethyl .alpha.-bromoisobutyrate, propanoic acid, 2-bromo-2-methyl-, ethyl ester, 2-bromo-2-methylpropanoic acid, ethyl ester, ethyl-2-bromoisobutyrate, propanoic acid, bromo-2-methyl-, ethyl ester, ethyl 2-bromo-2-methyl-propanoate, propionic acid, 2-bromo-2-methyl-, ethyl ester
PubChem CID 11745
IUPAC Name ethyl 2-bromo-2-methylpropanoate
SMILES CCOC(=O)C(C)(C)Br

Specifications

Density 1.329
Boiling Point 162°C to 164°C
Flash Point 57°C (135°F)
Refractive Index 1.444
Quantity 50 g
UN Number UN2924
Beilstein 1098947
Solubility Information Soluble in alcohol,ether. Insoluble in water.
Formula Weight 195.06
Percent Purity ≥98%
Chemical Name or Material Ethyl 2-bromoisobutyrate
Show More Show Less
Hazard Category H226-H317-H318-H340
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P261-P272-P280g-P281-P303+P361+P353-P305+P351+P338-P308+P313-P310-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 209-980-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only