missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate, 95%, Thermo Scientific Chemicals

Catalog No. AAH5907603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5907603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5907603 Supplier Thermo Scientific Chemicals Supplier No. H5907603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 238749-50-3 | Molecular Formula: C9H8BrNO2S | Molecular Weight: 274.132 g/mol

Ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate is used as an important raw material in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate is used as an important raw material in organic synthesis.

Solubility
Sparingly soluble in water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 238749-50-3
Molecular Formula C9H8BrNO2S
Molecular Weight (g/mol) 274.132
MDL Number MFCD15730638
InChI Key KXGSKTSSCQBDOA-UHFFFAOYSA-N
Synonym ethyl 2-bromo-4h-thieno 3,2-b pyrrole-5-carboxylate, ethyl 2-bromothieno<3,2-b>pyrrole-5-carboxylate, 2-bromo-4h-thieno 3,2-b pyrrole-5-carboxylic acid ethyl ester, ethyl 2-bromo-4h-thieno 3.2-b pyrrole-5-carboxylate, 2-bromothieno 3,2-b-pyrrole-5-carboxylic acid ethyl ester, 2-bromothieno 3,2-b pyrrole-5-carboxylic acid ethyl ester
PubChem CID 13164349
IUPAC Name ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate
SMILES CCOC(=O)C1=CC2=C(N1)C=C(S2)Br

Specifications

Quantity 1 g
Beilstein 4426004
Solubility Information Sparingly soluble in water.
Formula Weight 274.13
Percent Purity 95%
Chemical Name or Material Ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only