missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Bromo-4-methylthiazole-5-carboxylate 97.0+%, TCI America™

Catalog No. E09451G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09451G 1 g
E09455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09451G Supplier TCI America Supplier No. E09451G
Only null left

Chemical Identifiers

CAS 22900-83-0
Molecular Formula C7H8BrNO2S
Molecular Weight (g/mol) 250.11
MDL Number MFCD03791227
InChI Key CFBIOWPDDZPIDP-UHFFFAOYSA-N
Synonym ethyl 2-bromo-4-methylthiazole-5-carboxylate, 5-thiazolecarboxylic acid, 2-bromo-4-methyl-, ethyl ester, 2-bromo-4-methyl-thiazole-5-carboxylic acid ethyl ester, ethyl 2-bromo-4-methyl-5-thiazolecarboxylate, 2-bromo-4-methyl-5-thiazolecarboxylic acid ethyl ester, 2-bromo-4-methylthiazole-5-carboxylic acid ethyl ester, maybridge3_004543, ksc207i5t, ethyl2-bromo-4-methyl-1,3-thiazole-5-carboxylate, ethylbromomethylthiazolecarboxylate
PubChem CID 2824057
IUPAC Name ethyl 2-bromo-4-methyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(N=C(S1)Br)C

Specifications

Melting Point 68°C
Quantity 1 g
Formula Weight 250.11
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-Bromo-4-methylthiazole-5-carboxylate