missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate, 96%

Catalog No. AAH3411603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3411603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3411603 Supplier Thermo Scientific Chemicals Supplier No. H3411603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 40106-13-6 | C12H17NO2S | 239.333 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in methanol and ethanol.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 40106-13-6
Molecular Formula C12H17NO2S
Molecular Weight (g/mol) 239.333
MDL Number MFCD00216932
InChI Key XTUHIGALMIGZST-UHFFFAOYSA-N
Synonym ethyl 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta b thiophene-3-carboxylate, ethyl 2-amino-4h,5h,6h,7h,8h-cyclohepta b thiophene-3-carboxylate, ethyl 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta-b thiophene-3-carboxylate, ethyl 2-amino-4,5,6,7,8-pentahydrocyclohepta 1,2-b thiophene-3-carboxylate, 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta b thiophene-3-carboxylic acid ethyl ester, ethyl 2-aminocyclohepta b thiophene-3-carboxylate, aronis24232, ethyl 2-aminocycloheptathiophene-3-carboxylate, 2-amino-3-ethoxy-carbonyl-4,5-pentamethylenethiophene
PubChem CID 292644
IUPAC Name ethyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC2=C1CCCCC2)N

Specifications

Melting Point 87°C to 88°C
Quantity 1 g
Solubility Information Soluble in methanol and ethanol.
Formula Weight 239.33
Percent Purity 96%
Chemical Name or Material Ethyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate
Hazard Category H302+H332
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only