missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Amino-4,5-dimethylthiophene-3-carboxylate 98.0+%, TCI America™

Catalog No. p-7129274
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10055G 5 g
E100525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10055G Supplier TCI America Supplier No. E10055G

Chemical Identifiers

CAS 4815-24-1
Molecular Formula C9H13NO2S
Molecular Weight (g/mol) 199.268
MDL Number MFCD00085051
InChI Key JYSDXODDWAQWJR-UHFFFAOYSA-N
Synonym ethyl 2-amino-4,5-dimethyl3-thenoate, ethyl 2-amino-4,5-dimethyl-3-thiophenecarboxylate, 2-amino-4,5-dimethylthiophene-3-carboxylic acid ethyl ester, 2-amino-4,5-dimethyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-4,5-dimethyl-, ethyl ester, 2-amino-3-carbethoxy-4,5-dimethylthiophene, 2-amino-4,5-dimethyl-3-ethoxycarbonyl-thiophene, ethyl-2-amino-4,5-dimethylthiophene-3-carboxylate, ethyl2-amino-4,5-dimethylthiophene-3-carboxylate, ethyl 2-amino-4,5-dimethyl-thiophene-3-carboxylate
PubChem CID 78536
IUPAC Name ethyl 2-amino-4,5-dimethylthiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC(=C1C)C)N

Specifications

Melting Point 93°C
Color Yellow
Quantity 5 g
Formula Weight 199.27
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-Amino-4,5-dimethylthiophene-3-carboxylate