missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-4-(4-pyridyl)thiophene-3-carboxylate, 97%, Thermo Scientific Chemicals

Catalog No. AAH5806903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5806903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5806903 Supplier Thermo Scientific Chemicals Supplier No. H5806903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 117516-88-8 | Molecular Formula: C12H12N2O2S | Molecular Weight: 248.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Incompatible with oxidizing agents. Store in a dry, cool conditions in a well sealed containers.

Chemical Identifiers

CAS 117516-88-8
Molecular Formula C12H12N2O2S
Molecular Weight (g/mol) 248.3
MDL Number MFCD01922021
InChI Key LKLAPQXKZAVVPT-UHFFFAOYSA-N
Synonym ethyl 2-amino-4-4-pyridinyl-3-thiophenecarboxylate, ethyl 2-amino-4-pyridin-4-yl thiophene-3-carboxylate, ethyl 2-amino-4-4-pyridyl thiophene-3-carboxylate, 2-amino-4-pyridin-4-yl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-4-4-pyridinyl-, ethyl ester, 2-amino-4-4-pyridyl thiophene-3-carboxylic acid ethyl ester, acmc-20mn8d, cbmicro_021712, ethyl 2-azanyl-4-pyridin-4-yl-thiophene-3-carboxylate, 2-amino-4-pyridin-4-yl-3-thiophenecarboxylic acid ethyl ester
PubChem CID 874726
IUPAC Name ethyl 2-amino-4-pyridin-4-ylthiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC=C1C2=CC=NC=C2)N

Specifications

Quantity 1 g
Formula Weight 248.29
Percent Purity 97%
Chemical Name or Material Ethyl 2-amino-4-(4-pyridyl)thiophene-3-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only