Learn More
Ethyl 2,3-dibromopropionate, 97%
CAS: 3674-13-3 | C5H8Br2O2 | 259.925 g/mol
Supplier: Thermo Scientific Chemicals A1559122
Description
Ethyl 2,3-dibromopropionate is used as pharmaceutical Intermediates. It is a building block that has been used as a catalyst for the polymerization of ethyl acrylate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 2,3-dibromopropionate is used as pharmaceutical Intermediates. It is a building block that has been used as a catalyst for the polymerization of ethyl acrylate.
Solubility
Insoluble in water.
Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent, acids, bases.
Specifications
| Ethyl 2,3-dibromopropionate | |
| 1.789 | |
| 90°C (194°F) | |
| C5H8Br2O2 | |
| MFCD00000212 | |
| UN2922 | |
| ethyl 2,3-dibromopropionate, propanoic acid, 2,3-dibromo-, ethyl ester, ethyl alpha,beta-dibromopropionate, propionic acid, 2,3-dibromo-, ethyl ester, ethyl-2,3-dibromopropionate, 2,3-dibromopropionic acid ethyl ester, ethyl .alpha.,.beta.-dibromopropionate, propionic acid, 2,3-dibromo-, ethyl ester 6ci,7ci,8ci, ethyl2,3-dibromopropionate, acmc-1ahn6 | |
| OENICUBCLXKLJQ-UHFFFAOYSA-N | |
| ethyl 2,3-dibromopropanoate | |
| 97945 | |
| 97% |
| 3674-13-3 | |
| 212°C to 214°C | |
| Irritating | |
| 1.5 00 | |
| 100 g | |
| 636046 | |
| Insoluble in water. | |
| CCOC(=O)C(CBr)Br | |
| 259.925 | |
| 259.94 |
Chemical Identifiers
| 3674-13-3 | |
| 259.925 | |
| OENICUBCLXKLJQ-UHFFFAOYSA-N | |
| ethyl 2,3-dibromopropanoate |
| C5H8Br2O2 | |
| MFCD00000212 | |
| 97945 | |
| CCOC(=O)C(CBr)Br |
Safety and Handling
GHS H Statement
H301-H311-H314-H318-H227
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H301-H311-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 222-941-8
RTECSNumber : UA2458382
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only