missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,3-dibromopropionate, 97%

Catalog No. p-7022699
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
AAA1559114 25 g
AAA1559122 100 g
AAA1559136 500 g
3 options

Catalog No. AAA1559114

Supplier: Thermo Scientific Chemicals A1559114

Only null left

CAS: 3674-13-3 | C5H8Br2O2 | 259.925 g/mol

Ethyl 2,3-dibromopropionate is used as pharmaceutical Intermediates. It is a building block that has been used as a catalyst for the polymerization of ethyl acrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2,3-dibromopropionate is used as pharmaceutical Intermediates. It is a building block that has been used as a catalyst for the polymerization of ethyl acrylate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent, acids, bases.

Chemical Identifiers

CAS 3674-13-3
Molecular Formula C5H8Br2O2
Molecular Weight (g/mol) 259.925
MDL Number MFCD00000212
InChI Key OENICUBCLXKLJQ-UHFFFAOYSA-N
PubChem CID 97945
IUPAC Name ethyl 2,3-dibromopropanoate
SMILES CCOC(=O)C(CBr)Br

Specifications

Density 1.789
Boiling Point 212°C to 214°C
Flash Point 90°C (194°F)
Odor Irritating
Refractive Index 1.5 00
Quantity 25 g
UN Number UN2922
Beilstein 636046
Synonym ethyl 2,3-dibromopropionate, propanoic acid, 2,3-dibromo-, ethyl ester, ethyl alpha,beta-dibromopropionate, propionic acid, 2,3-dibromo-, ethyl ester, ethyl-2,3-dibromopropionate, 2,3-dibromopropionic acid ethyl ester, ethyl .alpha.,.beta.-dibromopropionate, propionic acid, 2,3-dibromo-, ethyl ester 6ci,7ci,8ci, ethyl2,3-dibromopropionate, acmc-1ahn6
Solubility Information Insoluble in water.
Formula Weight 259.94
Percent Purity 97%
Chemical Name or Material Ethyl 2,3-dibromopropionate
Show More Show Less
Hazard Category H227-H301-H311-H314-H335
Hazard Statement GHS H Statement
H301-H311-H314-H318-H227
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 222-941-8
RTECSNumber UA2458382
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only