accessibility menu, dialog, popup

UserName

H3COOBrBr

CAS RN 3674-13-3

IUPAC Name: ethyl 2,3-dibromopropanoate
Synonyms: ethyl 2,3-dibromopropanoate
Molecular Weight (g/mol): 259.93
Molecular Formula: C5H8Br2O2
InChi Key: OENICUBCLXKLJQ-UHFFFAOYNA-N
SMILES: CCOC(=O)C(Br)CBr
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)