missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(2-Amino-4-thiazolyl)-2-oxoacetate 96.0+%, TCI America™

Catalog No. E11595G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11595G 5 g
E115925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11595G Supplier TCI America Supplier No. E11595G

Chemical Identifiers

CAS 64987-08-2
Molecular Formula C7H8N2O3S
Molecular Weight (g/mol) 200.212
MDL Number MFCD00010414
InChI Key XNVRKLCQBZTGNA-UHFFFAOYSA-N
Synonym 2-(2-Amino-4-thiazolyl)-2-oxoacetic Acid Ethyl Ester
PubChem CID 4356513
IUPAC Name ethyl 2-(2-amino-1,3-thiazol-4-yl)-2-oxoacetate
SMILES CCOC(=O)C(=O)C1=CSC(=N1)N

Specifications

Melting Point 146°C
Color Yellow
Quantity 5 g
Formula Weight 200.21
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-(2-Amino-4-thiazolyl)-2-oxoacetate