missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1-(hydroxymethyl)cyclobutanecarboxylate, 97%

Catalog No. AAH62566MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62566MD 250 mg
AAH6256603 1 g
AAH6256606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH62566MD Supplier Thermo Scientific Chemicals Supplier No. H62566MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1195-81-9 | C8H14O3 | 158.197 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1195-81-9
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD11707219
InChI Key PWMQFMMZBJUHID-UHFFFAOYSA-N
Synonym ethyl 1-hydroxymethyl cyclobutane-1-carboxylate, ethyl 1-hydroxymethylcyclobutanecarboxylate, ethyl 1-hydroxymethyl cyclobutanecarboxylate, cyclobutanecarboxylic acid, 1-hydroxymethyl-, ethyl ester, ethyl 1-hydroxymethyl-cyclobutanecarboxylate, 1-hydroxymethyl-cyclobutanecarboxylic acid ethyl ester
PubChem CID 15670506
IUPAC Name ethyl 1-(hydroxymethyl)cyclobutane-1-carboxylate
SMILES CCOC(=O)C1(CCC1)CO

Specifications

Quantity 250 mg
Formula Weight 158.19
Percent Purity 97%
Chemical Name or Material Ethyl 1-(hydroxymethyl)cyclobutanecarboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only