missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 1,3-Dithiolane-2-carboxylate 97.0+%, TCI America™

Catalog No. D21055G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D21055G Supplier TCI America Supplier No. D21055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20461-99-8
Molecular Formula C6H10O2S2
Molecular Weight (g/mol) 178.264
MDL Number MFCD00005411
InChI Key OMCSHTHLIQOHDD-UHFFFAOYSA-N
Synonym 2-carboethoxydithiolane, ethyl1,3-dithiolane-2-carboxylate, 1,3-dithiolane-2-carboxylic acid ethyl ester, acmc-209faj, 2-carboethoxy-1,3-dithiolane, omcshthliqohdd-uhfffaoysa, 1,3-dithiolane-2-carboxylicacid,ethyl ester, 1,3-dithiolane-2-carboxylicacid, ethyl ester, 1,3-dithiolane-2-carboxylic acid, ethyl ester, ethyl 1,3-dithiolane-2-carboxylate
PubChem CID 88551
IUPAC Name ethyl 1,3-dithiolane-2-carboxylate
SMILES CCOC(=O)C1SCCS1

Specifications

Boiling Point 85°C
Quantity 5 g
Formula Weight 178.26
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 1,3-Dithiolane-2-carboxylate