missing translation for 'onlineSavingsMsg'
Learn More

(Ethoxycarbonylmethyl)dimethylsulfonium bromide, 98%

Catalog No. AAL0026618
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0026618 50 g
AAL0026609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0026618 Supplier Thermo Scientific Chemicals Supplier No. L0026618

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5187-82-6 | C6H13BrO2S | 229.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5187-82-6
Molecular Formula C6H13BrO2S
Molecular Weight (g/mol) 229.132
MDL Number MFCD00830557
InChI Key JXFPTJYKYKVENJ-UHFFFAOYSA-M
Synonym ethoxycarbonylmethyl dimethylsulfonium bromide, 2-ethoxy-2-oxoethyl dimethylsulfonium bromide, carbethoxymethyl-dimethylsulfonium bromide, 2-ethoxy-2-oxoethyl dimethylsulfanium bromide, 2-ethoxy-2-oxoethyl dimethyl sulfonium bromide, 2-ethoxy-2-oxoethyl dimethylsulphonium bromide, sulfonium, carbethoxymethyl dimethyl-, bromide, sulfonium, 2-ethoxy-2-oxoethyl dimethyl-, bromide, carbethoxymethyl dimethylsulfonium bromide, acmc-1az78
PubChem CID 78861
IUPAC Name (2-ethoxy-2-oxoethyl)-dimethylsulfanium;bromide
SMILES CCOC(=O)C[S+](C)C.[Br-]

Specifications

Melting Point ∽80°C (decomposition)
Quantity 50 g
Beilstein 4161472
Sensitivity Hygroscopic
Formula Weight 229.14
Percent Purity 98%
Chemical Name or Material (Ethoxycarbonylmethyl)dimethylsulfonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-968-3
RTECSNumber WR7600000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only