missing translation for 'onlineSavingsMsg'
Learn More

Ethoxyacetic Acid 98.0+%, TCI America™

Catalog No. E0377500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0377500G 500 g
E037725G 25 g
E0377100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E0377500G Supplier TCI America Supplier No. E0377500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 627-03-2
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.105
MDL Number MFCD00004310
InChI Key YZGQDNOIGFBYKF-UHFFFAOYSA-N
Synonym ethoxyacetic acid, acetic acid, ethoxy, ethoxy acetic acid, acetic acid, 2-ethoxy, ethoxy-acetic acid, unii-467vw095bx, o-ethylglycolic acid, ccris 7194, 2-ethoxy-acetic acid, dsstox_cid_11294
PubChem CID 12301
IUPAC Name 2-ethoxyacetic acid
SMILES CCOCC(=O)O

Specifications

Color Yellow
Boiling Point 207°C
Quantity 500 g
UN Number 3265
Formula Weight 104.11
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethoxyacetic Acid
EINECSNumber (2)-3131
RTECSNumber AH3600000
TSCA Yes