missing translation for 'onlineSavingsMsg'
Learn More

Ethidium bromide, 95%

Catalog No. AAL0748206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0748206 5 g
AAL0748203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0748206 Supplier Thermo Scientific Chemicals Supplier No. L0748206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1239-45-8 | C21H20BrN3 | 394.32 g/mol

For DNA isolation and visualization

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1239-45-8
Molecular Formula C21H20BrN3
Molecular Weight (g/mol) 394.32
MDL Number MFCD00011724
InChI Key ZMMJGEGLRURXTF-UHFFFAOYSA-N
Synonym ethidium bromide, homidium bromide, dromilac, 3,8-diamino-5-ethyl-6-phenylphenanthridinium bromide, ethydium bromide, 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide, etbr, 2,7-diamino-10-ethyl-9-phenylphenanthridinium bromide, 2,7-diamino-9-phenyl-10-ethylphenanthridinium bromide, unii-059nuo2z1l
PubChem CID 14710
ChEBI CHEBI:4883
SMILES [Br-].CC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12

Specifications

Melting Point ∽261°C (decomposition)
Odor Weak
Quantity 5 g
UN Number UN2811
Beilstein 3642536
Merck Index 14,4731
IUPAC Name 3,8-diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
Formula Weight 394.32
Percent Purity 98%
Assay (dry wt.)
Chemical Name or Material Ethidium bromide
Show More Show Less
Hazard Category H302-H330-H341
Hazard Statement GHS H Statement
H330-H341-H302
Fatal if inhaled.
Suspected of causing genetic defects.
Harmful if swallowed.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P301+P312-P304+P340-P308+P313-P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 214-984-6
RTECSNumber SF7950000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only