missing translation for 'onlineSavingsMsg'
Learn More

Ethanolamine, ACS reagent

Catalog No. AC451760025
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC451760025 2.5 L
AC451762500 250 mL
AC451760010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC451760025 Supplier Thermo Scientific Chemicals Supplier No. 451760025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 141-43-5 | C2H7NO | 61.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 141-43-5
Molecular Formula C2H7NO
Molecular Weight (g/mol) 61.08
MDL Number MFCD00008183
InChI Key HZAXFHJVJLSVMW-UHFFFAOYSA-N
Synonym ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine
PubChem CID 700
ChEBI CHEBI:16000
SMILES NCCO

Specifications

Melting Point 10.3°C
Density 1.0120g/mL
Boiling Point 170.0°C
Flash Point 92°C
Infrared Spectrum Authentic
Refractive Index 1.4530 to 1.4550
Linear Formula H2NCH2CH2OH
Quantity 2.5 L
Merck Index 15,3782
Specific Gravity 1.012
Solubility Information Solubility in water: soluble. Other solubilities: miscible with methanol and acetone, soluble in benzene, ether, ccl4 and n-heptane
IUPAC Name 2-aminoethanol
Viscosity 24 mPa.s (20°C)
Formula Weight 61.08
Percent Purity ≥99.0% (GC)
Grade ACS Reagent
Chemical Name or Material Ethanolamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED:
EINECSNumber 205-483-3

RUO – Research Use Only