missing translation for 'onlineSavingsMsg'
Learn More

Ethanesulfonic Acid 90.0+%, TCI America™

Catalog No. E003325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E003325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E003325G Supplier TCI America Supplier No. E003325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 594-45-6
Molecular Formula C2H5O3S
Molecular Weight (g/mol) 109.12
MDL Number MFCD00007529
InChI Key CCIVGXIOQKPBKL-UHFFFAOYSA-M
Synonym ethylsulfonic acid, ethanesulphonic acid, ethane sulfonate, ethane sulfonic acid, ethane sulphonic acid, ethansulfonic acid, ethylsulphonic acid, 1-ethanesulfonic acid, 2-ethanesulfonic acid, ethanesulfonicacid
PubChem CID 11668
ChEBI CHEBI:42465
IUPAC Name ethanesulfonate
SMILES CCS([O-])(=O)=O

Specifications

Melting Point -17°C
pH <1
Boiling Point 140°C
Quantity 25 g
UN Number 2586
Formula Weight 110.13
Percent Purity ≥90.0% (T)
Physical Form Liquid
Chemical Name or Material Ethanesulfonic Acid