missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Estriol, 97%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécifications
Spécifications
| Nom ou substance chimique | Estriol |
| CAS | 50-27-1 |
| Perte à la dessication | 0.5% max. |
| Spectre infrarouge | Authentic |
| Fourchette de pourcentages d’essai | 96% min. (HPLC) |
| Formule moléculaire | C18H24O3 |
| Quantité | 5g |
| Synonyme | estriol, oestriol, estratriol, ovestin, ovestrion, trihydroxyestrin, destriol, tridestrin, aacifemine, oestratriol |
| Renseignements sur la solubilité | Solubility in water: insoluble. Other solubilities: soluble in methanol |
| Clé InChI | PROQIPRRNZUXQM-ZXXIGWHRSA-N |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Suspected of causing cancer. May damage fertility or the unborn child. |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. Use personal protective equipment as required. IF exposed or concerned: Get medical advice/attention. |
| EINECSNumber | 200-022-2 |
RUO – Research Use Only