missing translation for 'onlineSavingsMsg'
Learn More

Estradiol Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111012206
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012206 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012206 Supplier Merck Emd Millipore Supplier No. PHR13531G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Estradiol is a steroid hormone present naturally in the human body. The availability of this hormone is quintessential in older women as it is a determinant of menopausal syndromes. It is utilized for the prevention of cardiovascular diseases.

Specifications

CAS 50-28-2
Physical Form Neat
Linear Formula C18H24O2
MDL Number MFCD00003693
Melting Point 176°C to 180°C
Synonym beta-Estradiol; 1,3,5-Estratriene-3,17 beta-diol; 17 beta-Estradiol; 3,17 beta-Dihydroxy-1,3,5(10)-estratriene; Dihydrofolliculin
Quantity 1 g
Formula Weight 272.38
Grade Certified Reference Material