missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Estradiene dione

Catalog No. AC463770050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463770050 5 g
AC463770010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC463770050 Supplier Thermo Scientific Chemicals Supplier No. 463770050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5571-36-8
Molecular Formula C20H26O3
Molecular Weight (g/mol) 314.43
InChI Key XUOQKQRMICQUQC-UHFFFAOYNA-N
IUPAC Name 11a-methyl-1,2,3,3a,3b,4,5,6,8,9,11,11a-dodecahydrospiro[cyclopenta[a]phenanthrene-7,2'-[1,3]dioxolan]-1-one
SMILES CC12CC=C3C(CCC4=C3CCC3(C4)OCCO3)C1CCC2=O

Specifications

Melting Point 156.0°C
Quantity 5 g
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in hot ethanol
Formula Weight 314.43
Chemical Name or Material Estradiene dione
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement May damage fertility.

May cause damage to organs through prolonged or repeated exposure.

Toxic to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only