missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Escitalopram oxalate

Catalog No. 479610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
479610 10 mg
479650 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 479610 Supplier Tocris Bioscience™ Supplier No. 4796/10

Selective serotonin reuptake inhibitor (SSRI)

Escitalopram oxalate is a selective 5-HT reuptake inhibitor (Ki values are 0.89, 8150, and 10500 nM for serotonin, noradrenalin, and dopamine transporters respectively). Escitalopram oxalate is an S-enantiomer and eutomer of citalopram (Cat. No. 1427).

Chemical Identifiers

CAS 219861-08-2
Molecular Formula C22H23FN2O5

Specifications

Quantity 10 mg
Formula Weight 414.43
Purity 98%
Molecular Weight (g/mol) 414.433
Chemical Name or Material Escitalopram oxalate
Recommended Storage Store at +4°C