missing translation for 'onlineSavingsMsg'
Learn More

Erythrosine B 95.0+%, TCI America™

Catalog No. T055725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T055725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T055725G Supplier TCI America Supplier No. T055725G

Chemical Identifiers

CAS 16423-68-0
Molecular Formula C20H6I4Na2O5
Molecular Weight (g/mol) 879.86
MDL Number MFCD00144257
InChI Key IINNWAYUJNWZRM-UHFFFAOYSA-L
Synonym Iodoeosine Sodium Salt, Tetraiodofluorescein Sodium Salt
PubChem CID 12961638
IUPAC Name disodium 2-(2,4,5,7-tetraiodo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
SMILES [Na+].[Na+].[O-]C(=O)C1=C(C=CC=C1)C1=C2C=C(I)C(=O)C(I)=C2OC2=C(I)C([O-])=C(I)C=C12

Specifications

Formula Weight 879.86
Percent Purity ≥95.0% (E)
Quantity 25 g
Physical Form Crystal/Powder
Chemical Name or Material Erythrosine B
EINECSNumber (5)-1503&(5)-1596
RTECSNumber LM5950000
TSCA Yes