missing translation for 'onlineSavingsMsg'
Learn More

Erythrosin B

Catalog No. AAA1418036
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1418036 500 g
AAA1418014 25 g
AAA1418022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1418036 Supplier Thermo Scientific Chemicals Supplier No. A1418036

Erythrosin B, C40H14I8Na2O10, CAS Number-16423-68-0, C.I. 45430; Tetraiodofluorescein sodium salt, 500g, 240-046-0, 879.87, H302, GHS H Statement H302 Harmful if swallowed., P264b-P270-P301+P312-P330-P501c, DKRYETBOLGDIOK-UHFFFAOYSA-L | CAS: 16423-68-0 | C40H14I8Na2O10 | 1715.757 g/mol

Erythrosin B, also called Acid Red 51, is used as a food colorant, in printing inks and as a biological stain in plant anatomy. It finds application as a dental plaque disclosing agent and a sensitizer for orthochromatic photographic films. As a food additive, it is commonly used in sweets like candies, popsicles and in cake-decorating gels. It is an important dye used in the determination of cell viability and used as phosphorescent triplet probe to detect rotational diffusion of membrane proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Erythrosin B, also called Acid Red 51, is used as a food colorant, in printing inks and as a biological stain in plant anatomy. It finds application as a dental plaque disclosing agent and a sensitizer for orthochromatic photographic films. As a food additive, it is commonly used in sweets like candies, popsicles and in cake-decorating gels. It is an important dye used in the determination of cell viability and used as phosphorescent triplet probe to detect rotational diffusion of membrane proteins.

Solubility
Soluble in water, glycerin and methanol.

Notes
Light sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place.Incompatible with oxidizing agents.

Chemical Identifiers

CAS 16423-68-0
MDL Number MFCD00144257
Synonym C.I. 45430; Tetraiodofluorescein sodium salt
PubChem CID 131676154

Specifications

Molecular Formula C40H14I8Na2O10
Solubility Information Soluble in water,glycerin and methanol.
InChI Key DKRYETBOLGDIOK-UHFFFAOYSA-L
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)I)O)I)I)I)C(=O)O.C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)I)[O-])I)I)I)C(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;2-(3-hydroxy-2,4,5,7-tetraiodo-6-oxoxanthen-9-yl)benzoic acid;2-(2,4,5,7-tetraiodo-3-oxido-6-oxoxanthen-9-yl)benzoate
Molecular Weight (g/mol) 1715.757
Formula Weight 879.87
Sensitivity Hygroscopic; Light sensitive
Quantity 500 g
Chemical Name or Material Erythrosin B
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 240-046-0
RTECSNumber LM5950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only