missing translation for 'onlineSavingsMsg'
Learn More

Erythrosin B, ∽98%, MP Biomedicals™

Catalog No. ICN19045005
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19045005 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19045005 Supplier MP Biomedicals Inc Supplier No. 0219045005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15905-32-5
Molecular Formula C20H8I4O5
Molecular Weight (g/mol) 835.90
MDL Number MFCD00005044
InChI Key OALHHIHQOFIMEF-UHFFFAOYSA-N
Synonym Solvent Red 140
PubChem CID 3259
IUPAC Name 3',6'-dihydroxy-2',4',5',7'-tetraiodo-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
SMILES OC1=C(I)C2=C(C=C1I)C1(OC(=O)C3=C1C=CC=C3)C1=CC(I)=C(O)C(I)=C1O2

Specifications

Assay Percent Range ∽98%
Color Red
Quantity 5 g
Physical Form Powder
Formula Weight 835.9
Chemical Name or Material Erythrosin B