missing translation for 'onlineSavingsMsg'
Learn More

Erythrosin B, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013299
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013299 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013299 Supplier Merck Emd Millipore Supplier No. 8761325MG
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 16423-68-0
Flash Point 182°C - closed cup
Physical Form Neat
Linear Formula C20H6I4 Na2O5
MDL Number MFCD00144257
Synonym Erythrosin extra bluish; 2′,4′,5′,7′-Tetraiodofluorescein disodium salt; Acid Red 51; Iodoeosin
Type Analytical Standard
Quantity 25 mg
Formula Weight 879.86
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Show More Show Less